Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

96085

Sigma-Aldrich

Z-Dap-OH

≥98.0% (NT)

Synonym(s):

Nα-Z-L-2,3-Diaminopropionic acid, Z-Dpr-OH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2CH(NHCO2CH2C6H5)CO2H
CAS Number:
Molecular Weight:
238.24
Beilstein:
4486950
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (NT)

form

crystals

optical activity

[α]20/D −39±1°, c = 1% in 1 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

mp

~240 °C (dec.)

application(s)

peptide synthesis

SMILES string

NC[C@H](NC(=O)OCc1ccccc1)C(O)=O

InChI

1S/C11H14N2O4/c12-6-9(10(14)15)13-11(16)17-7-8-4-2-1-3-5-8/h1-5,9H,6-7,12H2,(H,13,16)(H,14,15)/t9-/m0/s1

InChI key

FOXRXVSTFGNURG-VIFPVBQESA-N

Other Notes

Important building block for the synthesis of peptides containing DAP residues, e.g. bleomycins, edeines, tuberactinomycins

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C. Mendre et al.
Tetrahedron Letters, 35, 5429-5429 (1994)
S Shinagawa et al.
Journal of medicinal chemistry, 30(8), 1458-1463 (1987-08-01)
Emericedins A, B, and C, new betaines having inhibitory activity of long chain fatty acid oxidation, were isolated from the culture broth of Emericella quadrilineata IFO 5859. Their structures were determined by spectroscopic analyses as (R)-3-(acylamino)-4-(trimethylammonio)butyrate (acyl: A, acetyl; B
Y. Funabashi et al.
Tetrahedron, 49, 13-13 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service