Skip to Content
MilliporeSigma
All Photos(1)

Documents

96081

Sigma-Aldrich

N-Z-1,4-Butanediamine hydrochloride

≥98.0% (AT)

Synonym(s):

Benzyl N-(4-aminobutyl)carbamate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCONH(CH2)4NH2.HCl
CAS Number:
Molecular Weight:
258.74
Beilstein:
4725911
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (AT)

form

powder

reaction suitability

reagent type: cross-linking reagent

mp

192-196 °C

functional group

Cbz
amine

SMILES string

NCCCCNC(OCC1=CC=CC=C1)=O.[H]Cl

InChI

1S/C12H18N2O2.ClH/c13-8-4-5-9-14-12(15)16-10-11-6-2-1-3-7-11;/h1-3,6-7H,4-5,8-10,13H2,(H,14,15);1H

InChI key

ZVNNCIIFBSRHFE-UHFFFAOYSA-N

Other Notes

Protected diamine building block, e.g. synthesis of spermidine analogues

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J R Lakanen et al.
Journal of medicinal chemistry, 35(4), 724-734 (1992-02-21)
In order to assess the tolerance of the target enzyme spermine synthase for alpha-substituents on the aminopropyl moiety of the substrate spermidine, 1-methylspermidine (MeSpd, 2) was synthesized. It was determined that MeSpd is a poor substrate for spermine synthase and
I. Levchine et al.
Synthesis, 37-37 (1994)
A.G. Kalivretenos et al.
The Journal of Organic Chemistry, 58, 6596-6596 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service