Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

96060

Sigma-Aldrich

Z-Cha-OH

≥95.0% (HPLC)

Synonym(s):

Z-3-cyclohexyl-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23NO4
CAS Number:
Molecular Weight:
305.37
Beilstein:
5445123
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95.0% (HPLC)

optical activity

[α]20/D -13±2°, c = 1% in methanol

application(s)

peptide synthesis

SMILES string

OC(=O)[C@H](CC1CCCCC1)NC(=O)OCc2ccccc2

InChI

1S/C17H23NO4/c19-16(20)15(11-13-7-3-1-4-8-13)18-17(21)22-12-14-9-5-2-6-10-14/h2,5-6,9-10,13,15H,1,3-4,7-8,11-12H2,(H,18,21)(H,19,20)/t15-/m0/s1

InChI key

MNABZONTQXNLDT-HNNXBMFYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service