Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94647

Sigma-Aldrich

1-Boc-3-hydroxypiperidine

≥98.0% (TLC)

Synonym(s):

1-Boc-3-piperidinol, tert-Butyl 3-hydroxy-1-piperidinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19NO3
CAS Number:
Molecular Weight:
201.26
Beilstein:
4180859
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

description

remark: may darken during storage without loss in purity

Assay

≥98.0% (TLC)

form

crystals

SMILES string

CC(C)(C)OC(=O)N1CCCC(O)C1

InChI

1S/C10H19NO3/c1-10(2,3)14-9(13)11-6-4-5-8(12)7-11/h8,12H,4-7H2,1-3H3

InChI key

UIJXHKXIOCDSEB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service