Skip to Content
MilliporeSigma
All Photos(1)

Documents

94532

Sigma-Aldrich

1,4-Dioxa-8-azaspiro[4.5]decane hydrochloride

puriss., ≥99.5% (T)

Synonym(s):

4-Piperidone ethylene acetal hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO2 · HCl
CAS Number:
Molecular Weight:
179.64
Beilstein:
7304914
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

puriss.

Assay

≥99.5% (T)

form

crystals

mp

193-197 °C
193-198 °C

SMILES string

Cl[H].C1CC2(CCN1)OCCO2

InChI

1S/C7H13NO2.ClH/c1-3-8-4-2-7(1)9-5-6-10-7;/h8H,1-6H2;1H

InChI key

VXBPZOLCIITMPB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service