Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

944710343

Bis[2-(di-tert-butylphosphino)ethyl]amine solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H45NP2
Molecular Weight:
361.53
UNSPSC Code:
12352200
Pricing and availability is not currently available.

form

solution

Legal Information

Sold under license from Kanata for research purposes only. WO 2004096735.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service