Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94060

Sigma-Aldrich

Chloroacetic anhydride

technical, ≥90% (T)

Synonym(s):

Monochloroacetic acid anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClCH2CO)2O
CAS Number:
Molecular Weight:
170.98
Beilstein:
774533
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

<0.75 mmHg ( 20 °C)

grade

technical

Assay

≥90% (T)

color

brown

bp

120-123 °C/20 mmHg (lit.)

mp

48-60 °C (lit.)

SMILES string

ClCC(=O)OC(=O)CCl

InChI

1S/C4H4Cl2O3/c5-1-3(7)9-4(8)2-6/h1-2H2

InChI key

PNVPNXKRAUBJGW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

289.4 °F - closed cup

Flash Point(C)

143 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service