Skip to Content
MilliporeSigma
All Photos(1)

Documents

938858

Sigma-Aldrich

NanoFabTxPolymers for Microneedles

new

Polyvinyl alcohol

Synonym(s):

Poly(vinyl alcohol), Polyvinyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
12352104

description

80% hydrolyzed

Quality Level

form

crystals

falling ball

2.5-3.5 cP, 4 % in water(20 °C)

application(s)

advanced drug delivery
filtration

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

NanoFabTx™ Polymers for Microneedles are carefully selected polymers that can be used to formulate microneedle patches. Microneedles are tiny projections of polymeric needles that can vary in length, pitch, and array for topical drug delivery applications. They are typically composed of biodegradable synthetic or natural polymers at a variety of suitable concentrations.
Polyvinyl alcohol (PVA) is a synthetic water-soluble polymer and it widely used in many drug delivery applications. It is a biocompatible polymer which is available in different molecular weights. The formation of the microneedles depends on the molecular weight and concentration of PVA.
The NanoFabTx™ Polymers for Microneedles can be used with the microneedle templates to formulate microneedles that can be used for several applications, such as delivering various therapeutic molecules and vaccine antigens via the skin.
The selected polymer will enable users to:
1) save time in polymer selection by providing a starting point to formulate microneedles and avoiding lengthy trial-and-error optimization.
2)formulate microneedles for various drug delivery applications.

Legal Information

NANOFABTX is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microneedle arrays as transdermal and intradermal drug delivery systems: Materials science, manufacture and commercial development
Larra?eta E, et al.
Material Science and Engineering with Advanced Research, 104, 1-32 (2016)
Keegan Braz Gomes et al.
International journal of pharmaceutics, 613, 121393-121393 (2021-12-21)
Effective vaccines delivered via painless methods would revolutionize the way people approach vaccinations. This study focused on the development of fast-dissolving microneedles (MNs) to deliver antigen-loaded sustained release polymeric nanoparticles (NPs), achieving a dual-delivery platform for vaccination through the skin.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service