Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

938777

Cesium carbonate ChemBeads

new

Synonym(s):

Carbonic acid dicesium ChemBeads

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cs2CO3
Molecular Weight:
325.82
UNSPSC Code:
12352100

form

solid

Quality Level

pH

14-16(loading of base)

SMILES string

O=COO[Cs].[Cs]

InChI

1S/CH2O3.2Cs/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

FJDQFPXHSGXQBY-UHFFFAOYSA-L

General description

Cesium carbonate is a powerful inorganic base widely used in organic synthesis. It is a potential chemoselective catalyst for the reduction of aldehydes and ketones to alcohols. It is employed as base for the Heck coupling reaction of 4-trifluoromethyl-1-chlorobenzene and aryl chlorides.

Application

Cesium carbonate can be used as a base in C-C and C-N cross-coupling reactions such as Suzuki−Miyaura, Heck, and Buchwald-Hartwig amination reactions. It is also used as a base in alkylation reactions. Promotes the efficient O-alkylation of alcohols to form mixed alkyl carbonates.

For general uses, product is also available in powdered form(441902)

Features and Benefits

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

related product

Product No.
Description
Pricing

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - STOT RE 2 Oral

Target Organs

Kidney,Adrenal gland,Testes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service