Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

938246

Sigma-Aldrich

Silica

new

monodisperse, pore size (non-porous), 30 μm

Synonym(s):

Monodisperse silicon dioxide, Non-porous silica, Non-porous silicon dioxide, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352309

Quality Level

form

powder

particle size

30 μm

pore size

(non-porous)

bp

2230 °C (lit.)

mp

>1600 °C (lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our monodisperse, non-porous silica powder is a white powder composed of speherical particles, precisely engineered to achieve a monodisperse particle size of 30 µm. The microparticles do not contain any internal porosity or void spaces, making their surfaces impermeable to fluids or gases.

Application

Non-porous silica particles are widely recognized for their biocompatibility, making them a promising option for various biomedical applications. They have the potential to serve as carriers in drug delivery systems, imaging agents, and a bioconjugation platform for diagnostic assays. In the field of biomedicine, these particles can be employed to deliver drugs, proteins, and biomolecules through conjugation.[,] In the field of electronics, monodisperse non-porous silica can be self-assembled into arrays for optoelectronic applications like lenses, dielectric layers, and patterning at micron lengthscales.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Safety of Nonporous Silica Nanoparticles in Human Corneal Endothelial Cells
Kim, JY, et al.
Scientific Reports, 7(1), 14566-14566 (2017)
Core-shell particles lead the way to renewing high-performance liquid chromatography
Victor Gonzalez-Ruiz, et al.
TrAC, Trends in Analytical Chemistry, 64, 17-28 (2015)
Li Tang et al.
Nano today, 8(3), 290-312 (2013-09-03)
Nanomedicine, the use of nanotechnology for biomedical applications, has potential to change the landscape of the diagnosis and therapy of many diseases. In the past several decades, the advancement in nanotechnology and material science has resulted in a large number

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service