Skip to Content
MilliporeSigma
All Photos(1)

Documents

936359

Sigma-Aldrich

Chondroitin Sulfate A Methacrylate

15% methacrylated

Synonym(s):

CS-MA, CSMA, Chondroitin Sulfate, Chondroitin Sulfate A sodium salt Methacrylate, Chondroitin Sulfate Glycidyl Methacrylate, Chondroitin Sulfate acrylate, Chondroitin sulfate-methacrylate, Methacrylated chondroitin sulfate, Photocurable Chondroitin Sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C20H29N2NaO16S)n
UNSPSC Code:
12352202
NACRES:
NA.21

Quality Level

form

powder

color

white to light yellow, light brown

storage temp.

−20°C

General description

Chondroitin Sulfate A Methacrylate (CSMA) is a photo- and thermally crosslinkable biocompatible precursor for the preparation of hydrogels used in biomedical applications. Chondroitin Sulfate (CS) is a sulfated glycosaminoglycan (GAG) composed of a chain of alternating sugars, N-acetylgalactosamine and glucuronic acid, and is commonly found in the extracellular matrix (ECM) of connective tissues and cartilage. It is an important structural and mechanical components in tissues and its negatively charged sulfate group is known to interact electrostatically with positively charged growth factors to modulate signaling.

Application

  • Tissue engineering
  • 3D bioprinting
  • drug and gene delivery

Features and Benefits

  • Biocompatible
  • 15% methacrylated
  • Photo- or thermally crosslinkable
  • Negatively charged

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service