Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

936162

Sigma-Aldrich

RUMeIPr*

greener alternative

≥95%

Synonym(s):

2-(2,6-Dibenzhydryl-4-methylphenyl)-5-(dimethylamino)-2H-imidazo[1,5-a]pyridin-4-ium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H38ClN3
CAS Number:
Molecular Weight:
620.22
UNSPSC Code:
12161600
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥95%

form

powder

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

CN(C)C1=CC=CC2=CN(C3=C(C(C4=CC=CC=C4)C5=CC=CC=C5)C=C(C)C=C3C(C6=CC=CC=C6)C7=CC=CC=C7)C=[N+]21.[Cl-]

InChI

1S/C42H38N3.ClH/c1-31-27-37(40(32-17-8-4-9-18-32)33-19-10-5-11-20-33)42(44-29-36-25-16-26-39(43(2)3)45(36)30-44)38(28-31)41(34-21-12-6-13-22-34)35-23-14-7-15-24-35;/h4-30,40-41H,1-3H3;1H/q+1;/p-1

InChI key

PHYDGCNSIQWIAX-UHFFFAOYSA-M

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

RUMeIPr* is a NHC ligand developed by the Szostak Group. This NHC analogue of the priveledged DalPhos scaffold has been used in the following reactions: Au-catalyzed C-C coupling, Au-catalyzed C-N coupling, and Au-catalyzed hydroamination of internal alkynes.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service