Skip to Content
MilliporeSigma
All Photos(1)

Documents

936006

Sigma-Aldrich

Sodium bis(trifluoromethanesulfonyl)imide solution

new
greener alternative

in ethylene carbonate, dimethyl carbonate, 1.0 M NaTFSI in EC/DMC=50/50 (v/v), battery grade

Synonym(s):

1.0 M NaTFSI in EC/DMC electrolyte

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352600

grade

battery grade

Quality Level

form

solution

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

0.95-1.05 M (NaPF6)

impurities

≤100 ppm H2O

anion traces

chloride (Cl-): ≤200 ppm
sulfate (SO42-): ≤200 ppm

application(s)

battery manufacturing

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Sodium bis(trifluoromethanesulfonyl)imide solution in ethylene carbonate and dimethylcarbonate, 1.0 M NaTFSI in EC/DMC=50/50 (v/v) is a clear solution of battery grade sodium bis(trifluoromethanesulfonyl)imide (NaTfSI) dissolved in binary mixture of EC/DMC=50/50 (v/v).
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

The main application of 1.0 M NaTFSI in EC/DMC=50/50 (v/v), battery grade is as an electrolyte for sodium-ion batteries. NaTFSI-based electrolytes have advantages over NaPF6-based electrolytes because they are less sensitive to hydrolysis, safer to handle, and are more stable thermally and electrochemically. NaTFSI offers improved higher thermal stability compared to NaPF6 and NaBF4, and higher conductivity than NaTfO. The blend of ethylene carbonate and dimethyl carbonate achieves a low viscosity and good wettability of most polyolefin separators.[1,2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

77.0 °F

Flash Point(C)

25 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Non-aqueous electrolytes for sodium-ion batteries
A. Ponrouch, et al.
Journal of Material Chemistry A, 3, 22-42 (2015)
SEI Composition on Hard Carbon in Na-Ion Batteries After Long Cycling: Influence of Salts (NaPF6, NaTFSI) and Additives (FEC, DMCF)
J. Fondard ,et al.
Journal of the Electrochemical Society, 167 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service