Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

935794

Sigma-Aldrich

Multiphoton lithography bioink kit

gelatin-based hydrogel bioink

Synonym(s):

HYDROBIO INX X100

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NB.02

Quality Level

form

gel form

color

red

swelling

swells to 50-250 (Post processing total volume increase (%))

gel strength

(Storage modulus 4-26 kPa)

Looking for similar products? Visit Product Comparison Guide

General description

This gelatin-based bioink kit for multiphoton lithography (MPL) combines the biological benefits of conventional gelatin-based materials with two-photon polymerization processability. It is based on gelatin type B derived from natural collagen which is modified with photo-crosslinkable functional groups enabling efficient multiphoton processing at 780 nm in the presence of the supplied crosslinker. The gelatin-based bioink can be processed at fast speeds, due to the high reactivity and fast curing, resulting in high cell viability. The bioink is also biodegradable and allows cells to remodel the envrionment and substitute the hydrogel with newly formed extracellular matrix over time. This ready-to-use kit contains a concentrated resin, dilution buffer, and crosslinker for 10 prints.

Application

A wide range of biological applications including:
  • 3D bioprinting
  • Tissue engineering
  • High resolution printing of complex geometries

Features and Benefits

  • Biocompatibility - Supports cell adhesion and proliferation
  • Reproducibility - Production under strict quality control
  • Biodegradability - Enables cellular remodelling of the printed matrix
  • Easy handling - Delivered in a ready-to-use kit containing a concentrated stock solution, dilution buffer, and crosslinker for 10 prints that can be printed in 10 min
  • Biolgoically relevant - includes the benefits of collagen derived GelMA

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiphoton Lithography as a Promising Tool for Biomedical Applications
Greant C, et al.
Advanced Functional Materials , 33, 2212641-2212641 (2023)
Olivier Guillaume et al.
Acta biomaterialia, 165, 72-85 (2022-03-16)
Since its inception, tissue engineering and regenerative medicine (TERM) has been relying on either scaffold-based or scaffold-free strategies. Recent reports outlined the possibility of a synergistic, convergence approach, referred to as the third TERM strategy, which could alleviate bottlenecks of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service