Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

935611

Sigma-Aldrich

4-Vinyl-1,3-dioxolan-2-one

greener alternative

≥99.9%, anhydrous, battery grade

Synonym(s):

VEC, Vinyl ethylene carbonate

Sign Into View Organizational & Contract Pricing

Select a Size

935611-50G
$200.00

$200.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
935611-50G
$200.00

About This Item

Empirical Formula (Hill Notation):
C5H6O3
CAS Number:
Molecular Weight:
114.10
MDL number:
UNSPSC Code:
12352005

$200.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

battery grade

Quality Level

description

Application: Battery manufacturing

Assay

≥99.9%

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤100 ppm H2O

refractive index

n20/D 1.45 (lit.)

bp

237 °C/733 mmHg (lit.)

density

1.188 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

greener alternative category

storage temp.

2-8°C

SMILES string

C=CC1COC(=O)O1

InChI

1S/C5H6O3/c1-2-4-3-7-5(6)8-4/h2,4H,1,3H2

InChI key

BJWMSGRKJIOCNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Vinyl-1,3-dioxolan-2-one (VEC) is a cyclic organic compound with the molecular formula C5H6O3. It is a clear, colorless liquid with a high boiling point and is miscible with carbonate and ether solvents used in lithium-ion batteries (LIBs). Our battery grade VEC is differentiated by its high purity and low trace moisture to help ensure consistent performance in battery applications.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Anhydrous, battery grade 4-vinyl-1,3-dioxolan-2-one (VEC) is commonly used as an electrolyte additive in lithium-ion batteries (LIBs) to improve battery performance. VEC helps to form a stable film on the anode surface called a solid electrolyte interface (SEI). The SEI acts as a barrier that prevents further reaction between the electrolyte and the anode and thereby enhances the cyclic stability of the battery. For example, the addition of 2 wt.% VEC to the electrolyte formulation enhanced cycling performance in experiments both with Li metal anodes[1] and graphite anodes[2] In the electrolyte formulation, often VEC is added to be 2-10 wt.% of the solvent blend; however, scientists have even used VEC as the sole solvent in a lithium-metal battery with great results including long cycle life over 1000 cycles and reduced flammability[3]

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

206.4 °F - closed cup

Flash Point(C)

96.9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vinyl ethylene carbonate as an electrolyte additive for high-voltage LiNi0.4Mn0.4Co0.2O2/graphite Li-ion batteries
Zuo X, et al.
Ionics, 22, 201-208 (2016)
Vinyl Ethylene Carbonate as an Effective SEI-Forming Additive in Carbonate-Based Electrolyte for Lithium-Metal Anodes
Yang Y, et al.
ACS Applied Materials & Interfaces, 11, 6118-6125 (2019)
Highly safe and cyclable Li-metal batteries with vinylethylene carbonate electrolyte
Zhang Q, et al.
Nano Energy, 74, 104860-104860 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service