Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

935441

Sigma-Aldrich

Aldehyde functionalized hyaluronic acid

Degree of functionalization ~20%

Synonym(s):

HA-A, Hyaluronic acid, Hyluronic acid aldehyde, Oxidized hyaluronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$377.00

$377.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$377.00

About This Item

Linear Formula:
(C14H20NNaO11)n(C14H18NNaO11)m
UNSPSC Code:
12352201
NACRES:
NA.28

$377.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

powder or chunks (or fibers)

color

white to off-white

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Hyaluronic acid (HA) is an important component of the natural extracellular matrix which plays a vital role in various cellular and tissue functions[1]. It represents some unique properties such as excellent gel-forming properties and chemical modification ability through hydroxyl and carboxyl groups that make them potentially useful for their application in Tissue engineering[2]. Partially oxidized hyaluronate (OHA) containing two highly aldehyde groups in the repeating unit of HA. The dialdehyde groups of OHA are capable of reductive coupling with primary amines and thus can be used in a wide range of chemical modifications of HA as hydrogel for tissue engineering applications[3][4] and regenerative medicine[5][6] drug delivery applications[7][8]

Application

HA is increasingly used for a many biomedical applications because it promotes cell migration, angiogenesis, cell viability, and proliferation. Aldehyde functionalized HA can be used to develop hydrogels for tissue engineering, regenerative medicing, bioprinting, and drug delivery applications.

Features and Benefits

  • Coupling capabilities with primary amines
  • Biocompatible functionalized natural polymer
  • Promotes a variety of cellular and tissue functions

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Strategies for Hyaluronic Acid-Based Hydrogel Design in Drug Delivery
Sonia T, et al.
Pharmaceutics, 11, 407-407 (2019)
Oxi-HA/ADH Hydrogels: A Novel Approach in Tissue Engineering and Regenerative Medicine
Franca G C, et al.
Polysaccharides, 2, 477-496 (2021)
Modified hyaluronic acid-based materials for biomedical applications
Tiwari S, et al.
International Journal of Biological Macromolecules, 121, 556-571 (2019)
Hyaluronic Acid and Controlled Release: A Review
Bayer S I, et al.
Molecules (Basel), 25, 2649-2649 (2020)
General Dialdehyde Click Chemistry for Amine Bioconjugation
Elahipanah S, et al.
Bioconjugate Chemistry, 28, 1422-1433 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service