Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

934739

Sigma-Aldrich

Sodium manganese oxide

greener alternative

electrode sheet, aluminum substrate, size 5 in. × 10 in.

Synonym(s):

Manganese oxide cathode, Sodium-ion battery cathode

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$346.00

$346.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$346.00

About This Item

Empirical Formula (Hill Notation):
Na0.44MnO2
Molecular Weight:
97.05
UNSPSC Code:
26111700
NACRES:
NA.21

$346.00


Please contact Customer Service for Availability

Request a Bulk Order

material

aluminum substrate

Quality Level

grade

battery grade

description

2.5 V vs Na/Na+
Application: Battery Manufacturing

Assay

≥98%

form

sheet

composition

Active material loading 8.6 mg/cm2 ± 5%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

size

5 in. × 10 in.

thickness

16 μm , aluminum current collector
25-50 μm
72-80 μm , excluding current collector

particle size

1-2 μm (active material characteristic)

capacity

0.90 mAh/cm2±5 % (Areal capacity)

application(s)

battery manufacturing

greener alternative category

General description

Sodium manganese oxide (Na0.44MnO2) electrode sheet is a ready-to-use cathode for sodium-ion battery research. The film is 5 inches x 10 inches (127 mm x 254 mm) and cast single-sided on a 16 µm thick aluminum foil current collector. The composition is 90% sodium manganese oxide (Na0.44MnO2), 5% Poly(vinylidene fluoride) [PVDF] and 5% carbon black. The active material is tunnel-type sodium manganese oxide, which offers fast sodium ionic conductivity and high chemical stability. PVDF acts as binder due to its thermal and electrochemical stability and carbon black boosts the electronic conductivity.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

The primary application of our sodium manganese oxide (Na0.44MnO2) electrode sheet is as a cathode for sodium-ion battery research. Manganese-based compounds are widely investigated as cathode materials for rechargeable Na-ion batteries and, in particular, tunnel-type sodium manganese oxide (Na0.44MnO2) is a promising cathode material due to its high capacity, low cost, and non-toxicity. In addition, its unique tunnel structure facilitates excellent cycle and rate performance. For example, our sodium manganese oxide showed a large capacity (experimentally 110 mAh g−1 at 0.1C), even at fast charge rates (80 mAh g-1 at 5C, 72%). In addition, Na0.44MnO2 shows excellent stability with a variety of electrolytes including NaClO4 and NaPF6-based organic electrolytes.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrode Materials for Sodium-Ion Batteries
Kim H, et al.
Advanced Energy Materials, 6 (2016)
High-Performance Na0.44MnO2 Slabs for Sodium-Ion Batteries Obtained through Urea-Based Solution Combustion Synthesis
Ferrara C, et al.
Lithium-Ion Batteries, 4 (2018)
Xingde Xiang et al.
Advanced materials (Deerfield Beach, Fla.), 27(36), 5343-5364 (2015-08-15)
Sodium-ion batteries (SIBs) receive significant attention for electrochemical energy storage and conversion owing to their wide availability and the low cost of Na resources. However, SIBs face challenges of low specific energy, short cycling life, and insufficient specific power, owing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service