Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93433

Sigma-Aldrich

N,N′,N′′-Tritosyldiethylenetriamine

≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3C6H4SO2NHCH2CH2)2NSO2C6H4CH3
CAS Number:
Molecular Weight:
565.73
Beilstein:
1277399
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0%

form

powder

mp

172-176 °C (lit.)
172-176 °C

SMILES string

Cc1ccc(cc1)S(=O)(=O)NCCN(CCNS(=O)(=O)c2ccc(C)cc2)S(=O)(=O)c3ccc(C)cc3

InChI

1S/C25H31N3O6S3/c1-20-4-10-23(11-5-20)35(29,30)26-16-18-28(37(33,34)25-14-8-22(3)9-15-25)19-17-27-36(31,32)24-12-6-21(2)7-13-24/h4-15,26-27H,16-19H2,1-3H3

InChI key

OCDIAWYQMCPHAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Building block for the synthesis of polyaza macrocycles

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F. Chavez et al.
The Journal of Organic Chemistry, 54, 2990-2990 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service