Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

933341

Sigma-Aldrich

SEM Sample Holder

SEM Pin Stub Adapter, size 10 mm

Synonym(s):

stub adapter

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$183.00

$183.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$183.00

About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

$183.00


Please contact Customer Service for Availability

Request a Bulk Order

size

10 mm

Looking for similar products? Visit Product Comparison Guide

General description

EMS techware

Application

A simple to use adapter which will convert Hitachi, JEOL or ISI microscopes to use ⅛" pin stubs or Cambridge stubs.
Made of aluminum with spring retainers which will give a good connection.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service