Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93315

Sigma-Aldrich

Tris(tert-butyldimethylsilyl) phosphite

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H45O3PSi3
CAS Number:
Molecular Weight:
424.78
Beilstein:
4437247
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Product 93315 is not currently sold in your country. Contact Technical Service

Assay

≥97.0%

form

liquid

refractive index

n20/D 1.445

bp

82-84 °C/0.2 mmHg

mp

4-6 °C

density

0.907 g/mL at 20 °C (lit.)

SMILES string

CC(C)(C)[Si](C)(C)OP(O[Si](C)(C)C(C)(C)C)O[Si](C)(C)C(C)(C)C

InChI

1S/C18H45O3PSi3/c1-16(2,3)23(10,11)19-22(20-24(12,13)17(4,5)6)21-25(14,15)18(7,8)9/h1-15H3

InChI key

PEHSJBUJMHHQDE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent used for the preparation of phosphonic acid silyl esters from halides; the TBDMS esters are more stable than the corresponding TMS esters[1]; review[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.-L. Montero et al.
Tetrahedron Letters, 28, 1163-1163 (1987)
L. Wozniak et al.
Tetrahedron, 45, 2465-2465 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service