Skip to Content
MilliporeSigma
All Photos(1)

Documents

932957

Sigma-Aldrich

Tweezers Set

EMS DLC

Synonym(s):

Tweezer, Gripper

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

Looking for similar products? Visit Product Comparison Guide

General description

EMS techware

Application

Diamond Coated tweezer kit For fine and precision work: for demanding environments and working with hard samples or biological materials. Ideal for use in clean rooms. DLC tweezers tips are covered in a durable pure diamond film grown directly on the exposed metallic substrate. Body is non-magnetic, low carbon stainless steel. Set includes style 1, style 2A, style 3C, style 5, and style 7.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service