Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

932604

Sigma-Aldrich

Ge-OMe TPA

Synonym(s):

4,4′,4′′,4′′′-Germanetetrayltetrakis(N,N-bis(4-methoxyphenyl)aniline)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C80H72GeN4O8
Molecular Weight:
1290.09
UNSPSC Code:
12352103
NACRES:
NA.21
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

composition

Carbon content, 73-76%

storage temp.

2-8°C

SMILES string

COC(C=C1)=CC=C1N(C(C=C2)=CC=C2[Ge](C3=CC=C(N(C4=CC=C(OC)C=C4)C5=CC=C(OC)C=C5)C=C3)(C6=CC=C(N(C7=CC=C(OC)C=C7)C8=CC=C(OC)C=C8)C=C6)C9=CC=C(N(C%10=CC=C(OC)C=C%10)C%11=CC=C(OC)C=C%11)C=C9)C%12=CC=C(OC)C=C%12

InChI key

XBJLELSVFCNTIY-UHFFFAOYSA-N

Application

Organic hole transport layer (HTL) play very important role for realizing high performance and low-cost planar p-i-n perovskite solar cells (pero-SCs).Our 4,4′,4′′,4′′′-germanetetrayltetrakis(N,N-bis(4-methoxyphenyl)aniline), also known as Ge-OMe TPA can substantially simplify synthetic procedure by reducing device cost. This HTL can also facilitate the sequent growth of high-quality perovskite films, giving a significantly enhanced photovoltaic efficiency.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular design with silicon core: toward commercially available hole transport materials for high-performance planar p?i?n perovskite solar cells
Xue R, et al.
Journal of Material Chemistry A, 6, 404-413 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service