Skip to Content
MilliporeSigma
All Photos(2)

Documents

932167

Sigma-Aldrich

Poly[(9,9-dioctylfluorenyl-2,7-diyl)-co-(4,4′-(N-(p-butylphenyl))diphenylamine)]

Synonym(s):

Poly((4-butylphenyl)diphenylamine-alt-9,9-dioctylfluorene)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C51H61N)n
CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

mol wt

Mw ≥27,000 by GPC

Quality Level

solubility

chlorobenzene: soluble
chloroform: soluble
dichlorobenzene: soluble

λmax

434 nm±10 nm in THF

fluorescence

λem 385 nm±5 nm in THF

Application

Use as solution-processable Hole Transport / Hole Injection Layer (HTL / HIL) for organic electronics.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liangdong Zhang et al.
The journal of physical chemistry letters, 10(11), 3171-3175 (2019-05-24)
Two-dimensional (2D) perovskites incorporating hydrophobic organic spacer cations show improved film stability and morphology compared to their three-dimensional (3D) counterparts. However, 2D perovskites usually exhibit low photoluminescence quantum efficiency (PLQE) owing to strong exciton-phonon interaction at room temperature, which limits
Ling Chen et al.
ACS applied materials & interfaces, 10(28), 24232-24241 (2018-06-27)
Even though chemically stable metal oxides (MOs), as substitutes for poly(3,4-ethylenedioxythiophene):polystyrene sulfonate (PEDOT:PSS), have been successfully adopted for improving device stability in solution-processed quantum dot light-emitting diodes (QLEDs), the efficiencies of QLEDs are at a relatively low level. In this
Optimization of carrier transport layer: A simple but effective approach toward achieving high efficiency all-solution processed InP quantum dot light emitting diodes
Zhu X, et al.
Organic Electronics, 96, 106256-106256 (2021)
Inkjet printing a small-molecule binary emitting layer for organic light-emitting diodes.
Mu L, et al.
Journal of Material Chemistry C, 8, 6906-6913 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service