Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

931632

Sigma-Aldrich

Fmoc-L-H2PyrmOMe-OH

Novabiochem®

Synonym(s):

(αS)-α-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-5-methoxy-2-pyrimidinebutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H23N3O5
CAS Number:
Molecular Weight:
433.46
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

storage temp.

−20°C

Application

Fmoc-L-H2PyrmOMe-OH is an arginine analog that can be used to prepare novel peptides for usage as putative therapeutics or in the elucidation of structure-activity relationships.

Legal Information

Product of Peptidream
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service