Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

931586

Sigma-Aldrich

(P(t-Bu)3)Pd(4-CF3-C6H4)(Br)

greener alternative

Synonym(s):

Bromo[tris(1,1-dimethylethyl)phosphine][4-(trifluoromethyl)phenyl]palladium, P(t-Bu)3 Pd G6 Br

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$211.00
500 MG
$529.00

$211.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$211.00
500 MG
$529.00

About This Item

Empirical Formula (Hill Notation):
C19H31BrF3PPd
Molecular Weight:
533.74
UNSPSC Code:
12161600
NACRES:
NA.21

$211.00


Please contact Customer Service for Availability

Request a Bulk Order

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

Quality Level

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

2-8°C

SMILES string

Br[Pd]([P](C(C)(C)C)(C(C)(C)C)C(C)(C)C)C1=CC=C(C(F)(F)F)C=C1

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

(P(t-Bu)3)Pd(4-CF3-C6H4)(Br) is an oxidative addition complex that exhibits all of the same advantages of previous generations of Buchwald precatalysts.

Learn more about G6 Buchwald precatalysts

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service