Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

930865

Sigma-Aldrich

[CpNi(IPr)Cl]

greener alternative

≥95%

Synonym(s):

Cyclopentenylchloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]nickel

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$264.00

$264.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$264.00

About This Item

Empirical Formula (Hill Notation):
C32H41ClN2Ni
CAS Number:
Molecular Weight:
547.83
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.04

$264.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

−20°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

[CpNi(IPr)Cl] is an air and moisture stable Ni NHC catalyst for the Suzuki-Miyaura cross-coupling of amides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suzuki?Miyaura Cross-Coupling of Amides Using Well-Defined, Air- and Moisture-Stable Nickel/NHC (NHC = N-Heterocyclic Carbene) Complexes
Buchspies J., et al.
Catalysts, 10, 372-372 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service