929832
PCL-PEG-PCL diacrylate

average Mn (600-6000-600), poly(caprolactone)-b-poly(ethylene glycol)-b-poly(caprolactone) diacrylate
Synonym(s):
PCL-b-PEG-b-PCL, PEG hydrogel, acrylate-PCL-PEG-PCL-acrylate, biodegradable triblock copolymer
Select a Size
$650.00
Select a Size
About This Item
$650.00
Quality Level
form
low-melting solid
mol wt
average Mn (600-6000-600)
contains
contains ≤ 300 ppm MEHQ as inhibitor
color
white to off-white
Ω-end
acrylate
α-end
acrylate
storage temp.
−20°C
SMILES string
O=C(OCCOC(CCCCCOC(C=C)=O)=O)CCCCCOC(C=C)=O
InChI key
PCLZPCUCIICUMZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Tissue engineering
- 3D printing
- Drug delivery
Features and Benefits
- PCL-PEG-PCL triblock copolymer
- Hydrolytically degradable PCL blocks
- Acrylate functional groups permit photopolymerization
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service