Skip to Content
MilliporeSigma
All Photos(1)

Documents

929743

Sigma-Aldrich

[Ir(df(CF3)ppy)2(4,4′-(OMe)2bpy]BF4

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H22BF14IrN4O2
CAS Number:
Molecular Weight:
1011.59
UNSPSC Code:
12352101
NACRES:
NA.21

Quality Level

Assay

≥95%

form

powder or chunks

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

SMILES string

FC1=C(C2=CC(F)=C1)C3=[N](C=C(C=C3)C(F)(F)F)[Ir]452([N]6=CC=C(C=C6C7=CC(OC)=CC=[N]75)OC)[N]8=CC(C(F)(F)F)=CC=C8C9=C(C=C(C=C94)F)F.FB(F)(F)F

Application

[Ir(df(CF3)ppy)2(4,4′-(OMe)2bpy]BF4 is a versatile photocatalyst that can be used for a variety of synthetic transformations.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kennosuke Itoh et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(16), 5171-5179 (2020-12-11)
1,2,3,4-Tetrahydroquinolines should be applicable to the development of new pharmaceutical agents. A facile synthesis of 1,2,3,4-tetrahydroquinolines that is achieved by a photoinduced formal [4+2] cycloaddition reaction of acyclic α,β-unsaturated amides and imides with N,N-dialkylanilines under visible-light irradiation, in which a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service