Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

929646

Sigma-Aldrich

Poly(ethylene glycol) diacrylamide

average MN 10,000, acrylamide, MEHQ as stabilizer

Synonym(s):

PEG diacrylamide, PEG hydrogel, PEGDAAm, Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$512.00

$512.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$512.00

About This Item

Linear Formula:
H2CCHC(O)NH(CH2CH2O)n CH2CH2NHC(O)CHCH2
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.25

$512.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Poly(ethylene glycol) diacrylamide, Mn 10000, contains MEHQ as stabilizer

form

fine powder

Quality Level

mol wt

Mn 10000
average Mn 10000

contains

MEHQ as stabilizer

color

white to off-white

solubility

deionized water: 20 wt. % at 23 °C

Ω-end

acrylamide

α-end

acrylamide

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) diacrylamide is a linear, homobifunctional crosslinking PEG polymer with two acrylamide groups. PEG diacrylamide replaces the unstable ester bonds of commonly used PEG diacrylate crosslinkers with hydrolytically stable amides.

Application

PEG diacrylamide is a biocompatible, hydrophilic polymer commonly used to form crosslinked polymer networks and PEG hydrogels. Crosslinked networks can be formed via radical initiator induced polymerization and photopolymerization. Due to its stable amide bond, PEG diacrylamide can be used to prepare hydrogels suitable for long-term studies and eliminate non-specific hydrogel degradation. The acrylamide functional groups can also be reacted with thiol-containing peptide crosslinkers to create enzymatically-sensitive crosslinked networks. PEG diacrylamide has numerous applications in tissue engineering, 3D printing, and drug delivery.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service