Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

928291

Sigma-Aldrich

Platinum(IV) oxide ChemBeads

Synonym(s):

Adam’s catalyst, NSC 402624, Platinic oxide, Platinum dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$199.00

$199.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$199.00

About This Item

Linear Formula:
PtO2
CAS Number:
UNSPSC Code:
12352303
NACRES:
NA.21

$199.00


Please contact Customer Service for Availability

Request a Bulk Order

form

solid

Quality Level

extent of labeling

~5 wt. % loading of catalyst

SMILES string

[Pt]OO

InChI

1S/2O.Pt

InChI key

YKIOKAURTKXMSB-UHFFFAOYSA-N

Application

Platinum(IV) oxide has been used for the reduction of citernyl bromide to form (S)-3,7-imethyloctylbromide.Catalyst for hydrogenation of various functional groups with minimal hydrogenolysis problems; used as dehydrogenation catalyst; used for selective oxidation of primary alcohols.Catalyst for: Hydrohydrazination reactions, Oxidation of sugars, Low-temperature oxidation of carbon monoxide, Oxygen reduction reaction, Hydrogenation reactions, Hydrogenolysis of the cyclopropyl group into an isopropyl group, Hydroxycyclopropanation reactions.


ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

Learn more about ChemBeads products

For larger scale uses, product also available in powdered form (206032)

Pictograms

Flame over circleHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service