Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

928054

Sigma-Aldrich

Carbon nanotube, multi-walled

functionalized powder, L/D ~ 1000, ≥99.9 wt. % MWCNT basis

Synonym(s):

MWCNTs

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$123.00

$123.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$123.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NB.43

$123.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Multiwalled carbon nanotubes, functionalized powder, L/D ~ 1000, ≥99.9 wt. % MWCNT basis

Quality Level

Assay

≥99.9 wt. % MWCNT basis

composition

COOH Content, 1.20 wt. %

L

10-30 μm

diam.

10-20 nm , outer

surface area

≥233 m2/g

impurities

<0.1 wt. % Ash

conductivity

(Electrical Conductivity: >= 10-2S/cm)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Application

Our high-purity functionalized multiwalled carbon nanotubes can be used for nanocomposites to increase electrical conductivity or mechanical properties given its high conductivity and aspect ratio. Its functionalization allows stable formulations during your nanocomposite preparation. For instance, this product has shown stability within a gelatin based bioink: not observed any change in impedance over 6 months of real time and 3 years of accelerated time.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the electrical conductivity of 928054 carbon nanotube, multi-walled?

    1 answer
    1. The electrical conductivity of this product is >= 10^-2 S/cm.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service