Skip to Content
MilliporeSigma
All Photos(1)

Documents

928046

Sigma-Aldrich

Ready-to-Cast Slurry of MCMB (MesoCarbon MicroBeads) Graphite Powder for Lithium ion battery

Synonym(s):

Blade coatable ink slurry for Lithium ion battery

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352401
NACRES:
NA.23

Quality Level

Related Categories

Application

The liquid-type dispersion is made up of a common active material, a conductive agent, a binder and a solvent that researchers can readily use for processing lithium-ion battery electrodes.

Storage Class Code

10 - Combustible liquids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Effective Mixing for Lithium Ion Battery Slurries.
Liu D, et al.
Advances in Chemical Engineering and Science, 4, 515-528 (2014)
Conveying Advanced Li-ion Battery Materials into Practice The Impact of Electrode Slurry Preparation Skills.
Kraytsberg A, et al.
Advanced Energy Materials, 6, 1600655-1600655 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service