Skip to Content
MilliporeSigma
All Photos(1)

Documents

927694

Sigma-Aldrich

2-[6-(3-{4-[(tert-butoxy)carbonyl]piperazin-1-yl}propoxy)pyridin-3-yl]acetic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H29N3O5
Molecular Weight:
379.45
UNSPSC Code:
12352106
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

storage temp.

2-8°C

Application

2-[6-(3-{4-[(tert-butoxy)carbonyl]piperazin-1-yl}propoxy)pyridin-3-yl]acetic acid is a heterobifunctional crosslinker useful for development of PROTAC degraders for targeted protein degradation or other bifunctional molecules in chemical biology. Incorporation of rigidity into the linker region may impact the 3D orientation of the molecules and thus ternary complex formation as well as optimization of drug-like properties. For linkers already conjugated to E3 ligase ligands, please visit our collection of Protein Degrader Building Blocks.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service