Skip to Content
MilliporeSigma
All Photos(1)

Documents

925888

Sigma-Aldrich

6-(Dimethylamino)-9-(2,6-dimethylphenyl)-1-methoxy-10-phenylacridinium bromide

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H29BrN2O
CAS Number:
Molecular Weight:
513.47
UNSPSC Code:
12352101
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder or chunks

Application

6-(Dimethylamino)-9-(2,6-dimethylphenyl)-1-methoxy-10-phenylacridinium bromide is an acridinium photocatalyst used for a variety of transformations.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Legal Information

Product of Solvias

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bouthayna Zilate et al.
Chemical communications (Cambridge, England), 56(12), 1767-1775 (2020-01-31)
Recent developments in preparative photocatalysis have reshaped synthetic strategies and now represent an integral part of current organic chemistry. Due to their favourable electrochemical and photophysical properties, the nowadays most frequently used photocatalysts are based on precious Ru- and Ir-polypyridyl
Christian Fischer et al.
Angewandte Chemie (International ed. in English), 57(9), 2436-2440 (2017-12-19)
Despite the manifold use of heterocyclic fluorophores, only a fraction of the desired dye diversity can be accessed by contemporary synthetic approaches. Herein, we describe a modular method that converts various carboxylic acid esters directly into a broad spectrum of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service