Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

925799

Sigma-Aldrich

cis-(+)-carvomenthene oxide

Synonym(s):

7-Oxabicyclo[4.1.0]heptane, 1-methyl-4-(1-methylethyl)-, [1R-(1α,4α,6α)]-(9CI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
UNSPSC Code:
12352119
NACRES:
NA.22
Pricing and availability is not currently available.

form

liquid

Quality Level

functional group

ether

storage temp.

2-8°C

SMILES string

C[C@]1(O2)[C@@H]2C[C@H](C(C)C)CC1

InChI key

WSHVHJSDSVPPIV-KXUCPTDWSA-N

Application

cis-(+)-carvomenthene oxide is a valual chiral building block. Used in the synthesis of PSI(O), and generally for synthetic routes involving the addition of a reactive epoxide.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service