Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92461

Sigma-Aldrich

1,3,7-Trimethyluric acid

≥98.0% (HPLC)

Synonym(s):

2,6,8-Trihydroxy-1,3,7-trimethylpurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N4O3
CAS Number:
Molecular Weight:
210.19
Beilstein:
20503
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98.0% (HPLC)

mp

≥300 °C

SMILES string

CN1C(=O)N(C)C2=C(N(C)C(=O)N2)C1=O

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service