Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

921947

Sigma-Aldrich

N-Sulfinyl-O-(tert-butyl)hydroxylamine

≥95%

Synonym(s):

t-BuONSO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO2S
CAS Number:
Molecular Weight:
135.18
MDL number:
UNSPSC Code:
12352116
Pricing and availability is not currently available.

Quality Level

Assay

≥95%

form

liquid

refractive index

n/D 1.454

density

1.062 g/mL

storage temp.

−20°C

SMILES string

[S](=NOC(C)(C)C)=O

InChI

1S/C4H9NO2S/c1-4(2,3)7-5-8-6/h1-3H3

InChI key

QNDHWAYLAJVRCB-UHFFFAOYSA-N

Application

t-BuONSO is a stable reagent active for the synthesis of primary sulfonamides when used in conjunction with organolithium or Grignard reagents. t-BuONSO is tolerant of a variety of alkyl and aryl substrates and can be used in the synthesis of many medicinally relevant compounds.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Self-react. C - Skin Sens. 1

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Q Davies et al.
Journal of the American Chemical Society, 142(36), 15445-15453 (2020-08-26)
Sulfoximines and sulfonimidamides are promising compounds for medicinal and agrochemistry. As monoaza analogues of sulfones and sulfonamides, respectively, they combine good physicochemical properties, high stability, and the ability to build complexity from a three-dimensional core. However, a lack of quick
Thomas Q Davies et al.
Organic letters, 22(24), 9495-9499 (2020-11-26)
Sulfonamides have played a defining role in the history of drug development and continue to be prevalent today. In particular, primary sulfonamides are common in marketed drugs. Here we describe the direct synthesis of these valuable compounds from organometallic reagents
Ze-Xin Zhang et al.
Journal of the American Chemical Society, 141(33), 13022-13027 (2019-08-10)
Sulfondiimines-the double aza-analogues of sulfones-hold significant potential as leads in discovery chemistry, yet their application in this arena has been held back by the scarcity of appropriate synthetic routes. Existing methods employ sulfides as substrates, and rely on consecutive imination

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service