Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

919519

Sigma-Aldrich

Selenium nanoparticles

80 nm particle size, 0.15 wt. % in water, dispersion

Synonym(s):

SeNP

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$321.00

$321.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$321.00

About This Item

Empirical Formula (Hill Notation):
Se
CAS Number:
Molecular Weight:
78.96
MDL number:
UNSPSC Code:
12141910
NACRES:
NA.23

$321.00


Please contact Customer Service for Availability

Request a Bulk Order

form

dispersion

Quality Level

concentration

0.15 wt. % in water

particle size

80 nm

SMILES string

[Se]

InChI

1S/Se

InChI key

BUGBHKTXTAQXES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Selenium nanoparticles (SeNPs) are used in a wide variety of applications such as antimicrobial coatings, nanotherapeutics, diagnostics, and medical devices. These nanoparticles have been shown to play an important role in protecting against oxidative stress and have demonstrated low toxicity and bioavailability.[1][2]

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selenium Nanoparticles: Biomedical Applications.
Vrcek V, et al.
Springer Handbook of Nanotechnology (2018)
Amit Khurana et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 111, 802-812 (2019-01-08)
Nanoparticles (NPs) serve to reduce the toxicity, enhance bioactivity, improve targeting, and provide versatile means to control the release profile of the encapsulated moiety. Among different NPs, inorganic NPs of metals like Ag, Au, Ce, Fe, Se, Ti and Zn

Protocols

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service