Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

918857

Sigma-Aldrich

Tetracalcium phosphate (TTCP) powder

≥98%, ≤50 μm particle size

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$144.00

$144.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$144.00

About This Item

Linear Formula:
Ca4(PO4)2O
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

$144.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98%

form

powder

particle size

≤50 μm

SMILES string

[Ca].[P](=O)([O-])([O-])[O-].[O]

InChI

1S/Ca.H3O4P.O/c;1-5(2,3)4;/h;(H3,1,2,3,4);/p-3

InChI key

QOYIAXYUCQDEAW-UHFFFAOYSA-K

Application

Tetracalcium phosphate (TTCP) is used in calcium hydroxide cements, and hydrolyzes slowly to form calcium hydroxide and a thin insoluble apatite layer that prevents further reaction.[1] TTCP has a calcium phosphate phase with a Ca/P ratio greater than stoichiometric hydroxyapatite (HA) which makes it useful as a ceramic biomaterial.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcium phosphates in biomedical applications: materials for the future?
Habraken W, et al.
Materials Today, 19(2), 69-87 (2016)
Tetracalcium phosphate: Synthesis, properties and biomedical applications
Moseke C, et al.
Acta Biomaterialia, 6(10), 3815-3823 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service