Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91747

Sigma-Aldrich

Dimethylthexylsilyl trifluoromethanesulfonate

≥97.0% (T)

Synonym(s):

Dimethyl-(2,3-dimethyl-2-butyl)silyl trifluoromethanesulfonate, TDS triflate, Thexyl-dimethylsilyl trifluoromethanesulfonate, Trifluoromethanesulfonic acid thexyl-dimethylsilyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19F3O3SSi
CAS Number:
Molecular Weight:
292.39
Beilstein:
4186137
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

≥97.0% (T)

refractive index

n20/D 1.405

bp

53-54 °C/0.6 mmHg (lit.)

density

1.16 g/mL at 20 °C (lit.)

SMILES string

CC(C)C(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent used for the introduction of the thexyldimethylsilyl group; Review

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Simchen
Advances in Silicon Chemistry, 1, 189-189 null
D. Enders et al.
Angewandte Chemie (International Edition in English), 99, 359-359 (1987)
H.-J. Wetter et al.
Tetrahedron Letters, 26, 5515-5515 (1985)
T. Allmendinger et al.
Helvetica Chimica Acta, 71, 395-395 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service