Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

915718

Sigma-Aldrich

NanoFabTx reagent

PEGPLGA-75H

Synonym(s):

Formulation kit, NanoFabTx

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2·C2H4O)x·CH4O
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

application(s)

advanced drug delivery

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

NanoFabTx reagent, PEGPLGA-75H is a component of our NanoFabTx PEG-PLGA drug formulation screening kit (917796), which includes additional reagents and protocols for nanoprecipitation and microfluidics to synthesize 100-200 nm nanoparticles. NanoFabTx reagent, PEGPLGA-75H is a pegylated poly(lactic-co-glycolic acid) (PLGA) polymer. PLGA is a biocompatible and biodegradable polymer that has been used widely in drug delivery systems for controlled drug release of many different types of therapeutic molecules.

Application

NanoFabTx reagent, PEGPLGA-75H can be used as a refill for the component in our NanoFabTx PEG-PLGA drug formulation screening kit (917796).

Legal Information

NANOFABTX is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service