Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

914770

Sigma-Aldrich

Diethyl-4-((3r,5r,7r)-adamantan-1-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate

≥95%

Synonym(s):

4-Alkyl-1,4-dihydropyridine reagent, DHP reagent for light-mediated His modification

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H33NO4
CAS Number:
Molecular Weight:
387.51
UNSPSC Code:
12352200

Assay

≥95%

form

powder

storage temp.

−20°C

Application

Diethyl-4-((3r,5r,7r)-adamantan-1-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate was reported to chemoselectively modify histidine under visible light where the unsubstituted nitrogen groups on the modified His imidazole are conserved.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoping Chen et al.
Journal of the American Chemical Society, 141(45), 18230-18237 (2019-10-23)
Histidine (His) carries a unique heteroaromatic imidazole side chain and plays irreplaceable functional roles in peptides and proteins. Existing strategies for site-selective histidine modification predominantly rely on the N-substitution reactions of the moderately nucleophilic imidazole group, which inherently suffers from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service