Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

914010

Sigma-Aldrich

4-Nitrophenyl (2-(vinyloxy)ethyl) carbonate

≥95%

Synonym(s):

Light-initiated bioorthogonal photoclick cycloaddition reagent, VE, Vinyl ether photoclick reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO6
CAS Number:
Molecular Weight:
253.21
UNSPSC Code:
12352200

Assay

≥95%

form

liquid

storage temp.

−20°C

Application

4-Nitrophenyl (2-(vinyloxy)ethyl) carbonate is a vinyl ether photoclick reagent (VE) mediated by visible light for bioorthogonal reaction with a 9,10-phenanthrenequinone (PQ) photoclick reagent (see cat# 913766) for increased spatiotemporal control within biological applications. Signficantly, analysis of reactivity suggests this photoclick reaction is orthogonal to azide-alkyne and UV-mediated tetrazole-ene photoclick reactions, allowing for various means of simulteneous labeling of biomolecules in a given sample. This method requires no catalyst and is reported to be compatible with cells.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinbo Li et al.
Journal of the American Chemical Society, 140(44), 14542-14546 (2018-10-24)
Here we report a visible light-triggered, catalyst free bioorthogonal reaction that proceeds via a distinct pathway from reported bioorthogonal reactions. The prototype of this bioorthogonal reaction was the photocycloaddition of 9,10-phenanthrenequinone with electron-rich alkenes to form fluorogenic [4+2] cycloadducts. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service