Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

911941

Sigma-Aldrich

2-((Perfluorooctyl)sulfonyl)-1-phenylpropan-1-one

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$521.00

$521.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$521.00

About This Item

Empirical Formula (Hill Notation):
C17H9F17O3S
Molecular Weight:
616.29
UNSPSC Code:
12352101
NACRES:
NA.22

$521.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥95%

form

powder

reaction suitability

reaction type: C-C Bond Formation

mp

99-101 °C

Application

2-((Perfluorooctyl)sulfonyl)-1-phenylpropan-1-one is part of a suite of reagents developed by the Li group for late-stage functionalization of pharmaceutically relevant compounds. The reagents serve as precursors to alkyl radicals generated under light mediated, redox-neutral and catalyst free conditions for use in the Minisci alkylation of aromatics.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Lact. - Repr. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peng Liu et al.
Journal of the American Chemical Society, 139(40), 14315-14321 (2017-10-03)
The Minisci alkylation is useful to functionalize aromatics via alkyl radical addition. Current approaches to prepare alkyl radicals follow either oxidative or reductive pathways from various functional groups. Developing new strategy beyond these traditional methods remains elusive yet highly significant.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service