Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

911534

Sigma-Aldrich

(4,4′-dMeObpy)NiCl2

Synonym(s):

4,4′-dimethoxy-2,2′-bipyridine nickel (II) chloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$174.00
1 G
$542.00

$174.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$174.00
1 G
$542.00

About This Item

Empirical Formula (Hill Notation):
C12H12Cl2N2NiO2
CAS Number:
Molecular Weight:
345.84
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.23

$174.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder or crystals

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>300 °C

Application

(4,4′-dMeObpy)NiCl2 is a Ni catalyst used for a variety of cross-coupling reactions.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 Inhalation - Muta. 2 - Repr. 2 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhongdong Sun et al.
Organic letters, 19(14), 3727-3730 (2017-07-12)
Direct coupling of ethers and acyl halides was promoted by a binary catalytic system comprising an Ir-based photocatalyst and a Ni complex under blue-light irradiation. Photocatalysts with high triplet energy directed the catalysis, and the reaction likely proceeded by triplet-triplet

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service