Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

911135

Sigma-Aldrich

TU-3

Synonym(s):

4,4′-(2λ4δ2-benzo[1,2-c:4,5-c′]bis[1,2,5]thiadiazole-4,8-diyldi-5,2-thiophenediyl)bis[2-dodecylbenzonitrile], 4,8-Bis [5-(4-cyano-3-alkylphenyl)-2-thienyl] benzo[1,2-c:4,5-c′] bis [1,2,5] thiadiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H60N6S4
CAS Number:
Molecular Weight:
897.33
UNSPSC Code:
12352103
NACRES:
NA.23

description

Decomposition temperature: 371 °C (Decomposition start temperature)
mobility = 2.3 cm2 / Vs (@Vsd=100V)
on / off ratio = 107
Structure: BG-TC @ SiO2 / Si
electrode: Au
film: TU-3 / CHCl3 spin coat

Assay

≥99% (HPLC)

form

powder

mol wt

897.33 g/mol

color

dark green

mp

291 °C

storage temp.

2-8°C

Application

TU-3 is a n-type organic semiconductor material with long term (year-long) stability in air. And it can be deposited from its solutions to form felxible integrated circuits in either 2D or 3D configurations.
This material achieves a high electron mobility of 2.3 cm2/Vs or more in transistors, making it highly suitable for this application. The mobility of amorphous silicon used in general LCDs and other applications is about 0.5-1cm2/Vs.

Legal Information

Organic thin film transistor containing benzobis(thiadiazole) derivatives for organic electronic device by Tanaka, Yasuhiro; Kakita, Kazunari; Machida, Toshikazu From Jpn. Kokai Tokkyo Koho (2017), JP 2017079319 A 20170427.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasunori Takeda et al.
Scientific reports, 6, 25714-25714 (2016-05-10)
Ultrathin electronic circuits that can be manufactured by using conventional printing technologies are key elements necessary to realize wearable health sensors and next-generation flexible electronic devices. Due to their low level of power consumption, complementary (CMOS) circuits using both types
A Unique Solution-Processable n-Type Semiconductor Material Design for High-Performance Organic Field-Effect Transistors
Mamada, et al.
Chemistry of Materials, 27, 141-141 (2015)
Organic Complementary Inverter Circuits Fabricated with Reverse Offset Printing
Takeda Y, et al.
Advanced Electronic Materials, 4(1), 1700313-1700313 (2018)
Compact Organic Complementary D-Type Flip-Flop Circuits Fabricated with Inkjet Printing
Kazuma H, et al
Advanced Electronic Materials, 3(9), 1700208-1700208 (2019)
Jimin Kwon et al.
Nature communications, 10(1), 54-54 (2019-01-04)
Direct printing of thin-film transistors has enormous potential for ubiquitous and lightweight wearable electronic applications. However, advances in printed integrated circuits remain very rare. Here we present a three-dimensional (3D) integration approach to achieve technology scaling in printed transistor density

Articles

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Related Content

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service