Skip to Content
MilliporeSigma
All Photos(1)

Documents

908517

Sigma-Aldrich

Poly(D,L-lactide-b-glycolide) lactide:glycolide 50:50

amine terminated, average Mn 5,000

Synonym(s):

Amino-PLGA, Amino-functionalized PLGA, AminoPLGA, NH2 PLGA, PLGA-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H7N2(C3H4O2)x(C2H2O2)yH
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder or chunks

feed ratio

lactide:glycolide 50:50

mol wt

average Mn 5,000

color

white to tan

storage temp.

−20°C

Application

Biocompatible and biodegradable polymer can be used in the formation of nanoparticles for drug delivery. The amine functional group enables rapid and facile surface functionalization, allowing for this material to be used in applications such as targeted drug delivery.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yihan Xu et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 105(6), 1692-1716 (2016-04-22)
Poly (lactic-co-glycolic acid) (PLGA) copolymers have been broadly used in controlled drug release applications. Because these polymers are biodegradable, they provide an attractive option for drug delivery vehicles. There are a variety of material, processing, and physiological factors that impact
R Gref et al.
Science (New York, N.Y.), 263(5153), 1600-1603 (1994-03-18)
Injectable nanoparticulate carriers have important potential applications such as site-specific drug delivery or medical imaging. Conventional carriers, however, cannot generally be used because they are eliminated by the reticulo-endothelial system within seconds or minutes after intravenous injection. To address these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service