Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

907928

Sigma-Aldrich

(S)-2-((2-Hydroxy-3-(triphenylsilyl)benzyl)amino)-N,N,3-trimethylbutanamide

≥95%

Synonym(s):

Hoveyda aminophenol catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H36N2O2Si
CAS Number:
Molecular Weight:
508.73
UNSPSC Code:
12352111

Assay

≥95%

form

powder or crystals

mp

165-168 °C

storage temp.

2-8°C

Application

(S)-2-((2-Hydroxy-3-(triphenylsilyl)benzyl)amino)-N,N,3-trimethylbutanamide is an aminophenol organocatalyst developed by the Hoveyda lab for enantioselective addition of allylboron compounds to imines, ketones, and other carbonyls, including polyfluorinated substrates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Farid W van der Mei et al.
Journal of the American Chemical Society, 139(26), 9053-9065 (2017-06-27)
A practical method for enantioselective synthesis of fluoroalkyl-substituted Z-homoallylic tertiary alcohols has been developed. Reactions may be performed with ketones containing a polylfluoro-, trifluoro-, difluoro-, and monofluoroalkyl group along with an aryl, a heteroaryl, an alkenyl, an alkynyl, or an
KyungA Lee et al.
Nature chemistry, 8(8), 768-777 (2016-07-22)
Organofluorine compounds are central to modern chemistry, and broadly applicable transformations that generate them efficiently and enantioselectively are in much demand. Here we introduce efficient catalytic methods for the addition of allyl and allenyl organoboron reagents to fluorine-substituted ketones. These
Daniel W Robbins et al.
Angewandte Chemie (International ed. in English), 55(33), 9610-9614 (2016-06-09)
A set of broadly applicable methods for efficient catalytic additions of easy-to-handle allyl-B(pin) (pin=pinacolato) compounds to ketones and acyclic α-ketoesters was developed. Accordingly, a large array of tertiary alcohols can be obtained in 60 to >98 % yield and up to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service