Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

907316

Sigma-Aldrich

GlcA-ABP

95%

Synonym(s):

(2S,3S,4S,5R,6S)-6-(2-(difluoromethyl)-4-(hex-5-ynamido)phenoxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic acid, Glucuronic acid activity-based probe, Glucuronidated metabolite mimic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21F2NO8
Molecular Weight:
429.37
UNSPSC Code:
12352101

Assay

95%

form

solid

storage temp.

−20°C

Application

This glucuronic acid activity-based probe (GlcA-ABP) is a glucuronidated metabolite mimic that provides in situ detection of active β-glucuronidases in the gut microbiome. The probe reacts with nucleophilic residues of the target, and the alkyne tag enables subsequent bioorthogonal tagging via click chemistry for enrichment (i.e. biotin tags) or imaging and cell sorting (i.e. fluorescent tags). With GlcA-ABP, Whidbey, et al[1] were able to pair the activity-based probe (ABP) with fluorescence-activated cell sorting (FACS; ABP-FACS) to identify β-glucuronidase-active taxa in the gut microbiota.

Currently in gut microbiome research, access to and differentiation between subpopulations and their relationships to host biochemistry is a challenge. Functional and chemical approaches such as those with GlcA-ABP provide a complementary tool to gene sequencing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher Whidbey et al.
Journal of the American Chemical Society (2018-12-14)
Commensal microorganisms in the mammalian gut play important roles in host health and physiology, but a central challenge remains in achieving a de-tailed mechanistic understanding of specific microbial contributions to host biochemistry. New function-based approaches are needed that analyze gut

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service