Skip to Content
MilliporeSigma
All Photos(1)

Documents

906603

Sigma-Aldrich

DLAM-Iδ1Tγ-13CH3 Methyl Labeling Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.12

technique(s)

bio NMR: suitable

Quality Level

shipped in

dry ice

storage temp.

−70°C

Related Categories

General description

DLAM-Iδ1Tg-13CH3 kit has 13CH3isotopomer precursors and contains protocol instructions for creation of isotopically-labeled proteins.

Application

DLAM-Iδ1Tg-13CH3 is an optimized kit to produce proteins with 13CH3 isotopomer on the methyl groups of isoleucine and threonine. This kit has been tested with protein isotopic labeling in E. coli. It is used in methyl-TROSY NMR for the investigation of the active site of the proteasome and also in the study of catalytic mechanism.
For detection of long-range nOes or for the NMR study of large proteins

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Algirdas Velyvis et al.
Journal of the American Chemical Society, 135(25), 9259-9262 (2013-06-27)
The 20S proteasome core particle is a molecular machine that plays a central role in the regulation of cellular function through proteolysis, and it has emerged as a valuable drug target for certain classes of cancers. Central to the development
Silke Wiesner et al.
Current opinion in structural biology, 35, 60-67 (2015-09-26)
Intermolecular interactions are indispensible for biological function. Here we discuss how novel NMR techniques can provide unique insights into the assembly, dynamics and regulation of biomolecular complexes. We focus on applications that exploit the methyl TROSY effect and show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service